missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-phenylpropane, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-3-phenylpropane |
| CAS | 637-59-2 |
| Molecular Formula | C9H11Br |
| Quantity | 5 g |
| Synonym | (3-Bromopropyl)benzene, (gamma-Bromopropyl)benzene, 1-Bromo-3-phenylpropane, 1-Phenyl-3-bromopropane, 3-Bromo-1-phenylpropane, 3-Phenyl-1-bromopropane, 3-Phenylpropyl Bromide, NSC 133438 |
| InChI Formula | InChI=1S/C9H11Br/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2 |
| SMILES | BrCCCc1ccccc1 |
| IUPAC Name | 3-bromopropylbenzene |
| Molecular Weight (g/mol) | 199.09 |
| Formula Weight | 198.0044 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction