missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-phenylpropane, 98%

Product Code. 11497956
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497956 2500 g 2500g
11477956 100 g 100g
11487956 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497956 Supplier Thermo Scientific Alfa Aesar Supplier No. A14050.0E
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 637-59-2 | C9H11Br | 199.09 g/mol

1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-phenylpropane is used in the synthesis of MraY natural inhibitors as antibacterial agents. Also used in the preparation of quinolinone derivatives as potent and selective MAO-B inhibitors.

Solubility
Soluble in water (partly). Soluble in chloroform and methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-59-2
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.09
MDL Number MFCD00000257
InChI Key XMZQWZJMTBCUFT-UHFFFAOYSA-N
Synonym 3-bromopropyl benzene, 1-bromo-3-phenylpropane, 3-phenylpropyl bromide, benzene, 3-bromopropyl, 3-bromo-1-phenylpropane, 3-bromoprop-1-yl benzene, unii-6vx623qn9v, .gamma.-bromopropyl benzene, 4-bromopropylbenzene, benzene, bromopropyl
PubChem CID 12503
IUPAC Name 3-bromopropylbenzene
SMILES BrCCCC1=CC=CC=C1

Specifications

Density 1.308
Boiling Point 219°C to 220°C
Flash Point 101°C (213°F)
Refractive Index 1.546
Quantity 2500 g
Beilstein 2205527
Solubility Information Soluble in water (partly). Soluble in chloroform and methanol.
Formula Weight 199.09
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-phenylpropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.