missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-n-hexylbenzene, 97+%

Product Code. 11391999
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391999 25 g 25g
11381999 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391999 Supplier Thermo Scientific Alfa Aesar Supplier No. H26063.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38409-59-5 | C12H17Br | 241.172 g/mol

1-Bromo-3-n-hexylbenzene is used as intermediates in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-n-hexylbenzene is used as intermediates in pharmaceutical industry.

Solubility
Insoluble in water. Fully miscible with ethanol, acetone, toluene, methylene chloride.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38409-59-5
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.172
MDL Number MFCD09909567
InChI Key HZUVRCRPECDFAT-UHFFFAOYSA-N
Synonym 1-bromo-3-n-hexylbenzene, 1-bromo-3-hexyl-benzene, 3-hexylbromobenzene, 3-n-hexyl bromobenzene, #, benzene, 1-bromo-3-hexyl
PubChem CID 563022
IUPAC Name 1-bromo-3-hexylbenzene
SMILES CCCCCCC1=CC(=CC=C1)Br

Specifications

Density 1.179
Boiling Point 90°C to 98°C (5 mmHg)
Flash Point >120°C (248°F)
Refractive Index 1.522
Quantity 25 g
Solubility Information Insoluble in water. Fully miscible with ethanol,acetone,toluene,methylene chloride.
Formula Weight 241.17
Percent Purity ≥97%
Chemical Name or Material 1-Bromo-3-n-hexylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.