missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methylbutane, TRC

Product Code. 30136949
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136949 250 g 250g
30010161 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136949 Supplier Toronto Research Chemicals Supplier No. TRCB685555250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-82-4
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.04
Synonym 3-Methyl-1-bromobutane, 3-Methylbutyl Bromide, 4-Bromo-2-methylbutane, Isoamyl Bromide, Isobutylmethyl Bromide, Isopentyl Bromide, NSC 9240
IUPAC Name 1-bromo-3-methylbutane
SMILES CC(CCBr)C

Specifications

Quantity 250 g
InChI Formula InChI=1S/C5H11Br/c1-5(2)3-4-6/h5H,3-4H2,1-2H3
Formula Weight 150.0044
Chemical Name or Material 1-Bromo-3-methylbutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.