missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methylbutane, 99%

Product Code. 10540552 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10540552 5 g Glass bottle 5g
10607751 100 g Glass bottle 100g
10249450 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10540552 Supplier Thermo Scientific Acros Supplier No. 403050050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-82-4 | C5H11Br | 151.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-82-4
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.05
MDL Number MFCD00000253
InChI Key YXZFFTJAHVMMLF-UHFFFAOYSA-N
Synonym isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa
PubChem CID 7891
IUPAC Name 1-bromo-3-methylbutane
SMILES CC(C)CCBr

Specifications

Acidity 0.1% max. (as HBr)
CAS Min % 98.5
CAS Max % 100.0
Melting Point -112°C
Color Colorless
Density 1.2000g/mL
Boiling Point 120°C to 121°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH2Br
Refractive Index 1.4399 to 1.4419
Quantity 5 g
Beilstein 01, 136
Merck Index 15, 5159
Specific Gravity 1.2
Solubility Information Solubility in water: 0.2g/L in water (16°C). Other solubilities: miscible with alcohol,ether,benzene and,carbon tetrachloride
Formula Weight 151.05
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-methylbutane, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.