missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-iodobenzene, 98+%

Product Code. 10084623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10084623 50 mL Glass bottle 50mL
10536061 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10084623 Supplier Thermo Scientific Acros Supplier No. 249420500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-18-4 | C6H4BrI | 282.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-18-4
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.91
MDL Number MFCD00001043
InChI Key CTPUUDQIXKUAMO-UHFFFAOYSA-N
Synonym 3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
PubChem CID 11561
IUPAC Name 1-bromo-3-iodobenzene
SMILES BrC1=CC=CC(I)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -9.3°C
Density 2.2100g/mL
Boiling Point 120.0°C (18.0 mmHg)
Flash Point >110°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4I
Refractive Index 1.6590 to 1.6615
Quantity 50 mL
Beilstein 05, 223
Specific Gravity 2.21
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 282.89
Percent Purity 98+%
Chemical Name or Material 1-Bromo-3-iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-3-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.