missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-iodobenzene, 98%

Product Code. 11474147
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474147 25 g 25g
11484147 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474147 Supplier Thermo Scientific Alfa Aesar Supplier No. A19596.14
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 591-18-4 | C6H4BrI | 282.91 g/mol

1-Bromo-3-iodobenzene is used in the preparation of 3'-bromo-2,3,4,5,6-pentaethyl-biphenyl by reacting with hex-3-yne. It is also used to prepare 1-(3'-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene and 1-(3'-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene. Further, it is involved in the preparation of oxygen-tethered 1,6-enynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-iodobenzene is used in the preparation of 3′-bromo-2,3,4,5,6-pentaethyl-biphenyl by reacting with hex-3-yne. It is also used to prepare 1-(3′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene and 1-(3′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene. Further, it is involved in the preparation of oxygen-tethered 1,6-enynes.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-18-4
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.91
MDL Number MFCD00001043
InChI Key CTPUUDQIXKUAMO-UHFFFAOYSA-N
Synonym 3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
PubChem CID 11561
IUPAC Name 1-bromo-3-iodobenzene
SMILES BrC1=CC=CC(I)=C1

Specifications

Melting Point -9°C
Density 2.219
Boiling Point 252°C
Flash Point >110°C (230°F)
Odor Recognizable
Refractive Index 1.662
Quantity 25 g
Beilstein 1854384
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 282.91
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-iodobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.