missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-iodobenzene, 98+%, Thermo Scientific Chemicals

Catalog Number 10536061
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
591-18-4
C6H4BrI
282.91
MFCD00001043
CTPUUDQIXKUAMO-UHFFFAOYSA-N
3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
11561
1-bromo-3-iodobenzene
BrC1=CC=CC(I)=C1
This item is not returnable. View return policy
591-18-4
C6H4BrI
282.91
MFCD00001043
CTPUUDQIXKUAMO-UHFFFAOYSA-N
3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
11561
1-bromo-3-iodobenzene
BrC1=CC=CC(I)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -9.3°C
Density 2.2100g/mL
Boiling Point 120.0°C (18.0 mmHg)
Flash Point >110°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4I
Refractive Index 1.6590 to 1.6615
Quantity 10 mL
Beilstein 05, 223
Specific Gravity 2.21
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 282.89
Percent Purity 98+%
Chemical Name or Material 1-Bromo-3-iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-3-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'