missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-fluorobenzene, 99%

Product Code. 11419842
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419842 250 g 250g
11399206 10 g 10g
11464190 25 g 25g
11409842 50 g 50g
11474190 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11419842 Supplier Thermo Scientific Alfa Aesar Supplier No. A10858.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1073-06-9 | C6H4BrF | 175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-06-9
Molecular Formula C6H4BrF
MDL Number MFCD00000326
InChI Key QDFKKJYEIFBEFC-UHFFFAOYSA-N
Synonym 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
PubChem CID 14082
IUPAC Name 1-bromo-3-fluorobenzene

Specifications

Melting Point -8°C
Density 1.594
Boiling Point 148°C to 150°C
Flash Point 46°C (115°F)
Odor Aromatic
Refractive Index 1.5275
Quantity 250 g
UN Number UN1993
Beilstein 1853969
SMILES C1=CC(=CC(=C1)Br)F
Molecular Weight (g/mol) 175
Formula Weight 175.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-3-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.