missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3-ethylbenzene, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-3-ethylbenzene |
| CAS | 2725-82-8 |
| Molecular Formula | C8H9Br |
| Quantity | 25 g |
| Synonym | (3-Ethylphenyl)bromide, 3-Bromo-1-ethylbenzene, 3-Ethylbromobenzene, m-Bromoethylbenzene, m-Ethylbromobenzene |
| InChI Formula | InChI=1S/C8H9Br/c1-2-7-4-3-5-8(9)6-7/h3-6H,2H2,1H3 |
| SMILES | CCc1cccc(Br)c1 |
| IUPAC Name | 1-bromo-3-ethylbenzene |
| Molecular Weight (g/mol) | 185.06 |
| Formula Weight | 183.9888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction