missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-ethylbenzene, 97%, Thermo Scientific™

Product Code. p-2695625
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10150892

Brand: Thermo Scientific Maybridge TL00571DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2725-82-8
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00156128
InChI Key ZRFJYAZQMFCUIX-UHFFFAOYSA-N
Synonym benzene, 1-bromo-3-ethyl, 3-ethylbromobenzene, 1-bromo-3-ethyl-benzene, 3-bromoethyl benzene, 3ethylbromobenzene, m-bromoethylbenzene, 3-bromo-ethylbenzene, pubchem3771, 1bromo-3-ethylbenzene, 1-bromo-3-ethylbenze
PubChem CID 123170
IUPAC Name 1-bromo-3-ethylbenzene
SMILES CCC1=CC(=CC=C1)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CH3CH2C6H4Br
Quantity 1 g
Formula Weight 185.06
Percent Purity 97%
Chemical Name or Material 1-Bromo-3-ethylbenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.