missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-2-iodobenzene, 97%

Product Code. 15492679
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492679 1 g 1g
15412689 250 mg 250mg
15402689 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492679 Supplier Thermo Scientific Alfa Aesar Supplier No. H66020.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 450412-28-9 | C6H3BrClI | 317.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 450412-28-9
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.35
MDL Number MFCD09907935
InChI Key GIMVCZMZRZGDTL-UHFFFAOYSA-N
Synonym 2-bromo-6-chloroiodobenzene, 1-bromo-2-iodo-3-chlorobenzene, benzene, 1-?bromo-?3-?chloro-?2-?iodo
PubChem CID 10335963
IUPAC Name 1-bromo-3-chloro-2-iodobenzene
SMILES ClC1=C(I)C(Br)=CC=C1

Specifications

Quantity 1 g
Beilstein 9119447
Sensitivity Light sensitive
Formula Weight 317.35
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Bromo-3-chloro-2-iodobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.