missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-2-fluorobenzene, 95%

Product Code. 15488168
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15488168

Brand: Thermo Scientific Alfa Aesar H61622.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144584-65-6 | C6H3BrClF | 209.442 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144584-65-6
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD03412182
InChI Key DRNWNTAANHEQMK-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorobromobenzene, 1-bromo-2-fluoro-3-chlorobenzene, 1-bromo-3-chloro-2-fluoro-benzene, pubchem4338, acmc-1c0jw, intermediates-zcf02109, ksc493i6p, benzene,1-bromo-3-chloro-2-fluoro
PubChem CID 3685762
IUPAC Name 1-bromo-3-chloro-2-fluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)Cl

Specifications

Melting Point 42°C to 46°C
Color White to Yellow
Quantity 25 g
Formula Weight 209.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-Bromo-3-chloro-2-fluorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.