missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals

Product Code. 10397131 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10397131

Brand: Thermo Scientific 331900250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19752-55-7
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.9
MDL Number MFCD00000584
InChI Key DZHFFMWJXJBBRG-UHFFFAOYSA-N
Synonym 3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a
PubChem CID 29766
IUPAC Name 1-bromo-3,5-dichlorobenzene
SMILES C1=C(C=C(C=C1Cl)Br)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 74°C to 77°C
Boiling Point 232°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 225.9
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-3, 5-dichlorobenzene, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.