missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,4-dichlorobenzene, 98+%

Product Code. p-73882 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10499940

Brand: Thermo Scientific Acros 269810250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18282-59-2 | C6H3BrCl2 | 225.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-59-2
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.89
MDL Number MFCD00040849
InChI Key CFPZDVAZISWERM-UHFFFAOYSA-N
Synonym 1-bromo-3,4-dichlorobenzene, 3,4-dichlorobromobenzene, benzene, 4-bromo-1,2-dichloro, 3,4-dichlorophenyl bromide, 3,4-dichloro-1-bromobenzene, 4-bromo-1,2-dichloro-benzene, 5-bromdichlorbenzol, pubchem3611, acmc-1btol, 3,4-dichloro-bromobenzene
PubChem CID 29013
IUPAC Name 4-bromo-1,2-dichlorobenzene
SMILES ClC1=CC=C(Br)C=C1Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 23.0°C to 24.0°C
Color Yellow
Boiling Point 140.0°C to 142.0°C (30.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula BrC6H3Cl2
Refractive Index 1.5986 to 1.6006
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 225.91
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 1-Bromo-3, 4-dichlorobenzene, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-3,4-dichlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.