missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,3-dimethylbutan-2-one, Tech., Thermo Scientific™

Product Code. p-2695615
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10041794

Brand: Thermo Scientific Maybridge CD08380EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5469-26-1
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.057
MDL Number MFCD00000206
InChI Key SAIRZMWXVJEBMO-UHFFFAOYSA-N
Synonym 1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
PubChem CID 21642
IUPAC Name 1-bromo-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CBr

Specifications

CAS Min % 80.0
CAS Max % 100.0
Density 1.3300g/mL
Packaging Amber glass bottle
Linear Formula (CH3)3CCOCH2Br
Quantity 10 g
Formula Weight 179.06
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-Bromo-3, 3-dimethylbutan-2-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.