missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,3,4,4,4-pentafluoro-2-butanone, 97%, Thermo Scientific Chemicals

Catalog Number 11368665
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
92737-01-4
C4H2BrF5O
240.955
MFCD00041447
HHGIRNBWKHUFNA-UHFFFAOYSA-N
2736355
1-bromo-3,3,4,4,4-pentafluorobutan-2-one
C(C(=O)C(C(F)(F)F)(F)F)Br
This item is not returnable. View return policy
92737-01-4
C4H2BrF5O
240.955
MFCD00041447
HHGIRNBWKHUFNA-UHFFFAOYSA-N
2736355
1-bromo-3,3,4,4,4-pentafluorobutan-2-one
C(C(=O)C(C(F)(F)F)(F)F)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 93°C to 94°C
Quantity 1 g
UN Number UN3265
Synonym 1-bromo-3,3,4,4,4-pentafluoro-2-butanone, 2-butanone, 1-bromo-3,3,4,4,4-pentafluoro, 1-bromo-3,3,4,4,4-pentafluorobutanone, 2-butanone,1-bromo-3,3,4,4,4-pentafluoro, 1-bromanyl-3,3,4,4,4-pentakis fluoranyl butan-2-one
Formula Weight 240.96
Percent Purity 97%
Chemical Name or Material 1-Bromo-3,3,4,4,4-pentafluoro-2-butanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'