missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(trifluoromethylthio)benzene, 97%

Product Code. 11440050
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440050 5 g 5g
11430050 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440050 Supplier Thermo Scientific Alfa Aesar Supplier No. H50576.06
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1644-72-0 | C7H4BrF3S | 257.07 g/mol

1-Bromo-2-(trifluoromethylthio)benzene is used as organic chemical synthesis intermediate. It is also a useful halogenated compound for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-(trifluoromethylthio)benzene is used as organic chemical synthesis intermediate. It is also a useful halogenated compound for proteomics research.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1644-72-0
Molecular Formula C7H4BrF3S
Molecular Weight (g/mol) 257.07
MDL Number MFCD00236347
InChI Key IWRJVJJQWGZKMT-UHFFFAOYSA-N
Synonym 2-trifluoromethylthio bromobenzene, 1-bromo-2-trifluoromethylthio benzene, 1-bromo-2-trifluoromethyl sulfanyl benzene, 2-bromophenyl trifluoromethyl sulfane, 2-bromophenyl trifluoromethyl sulphide, pubchem20490, 1-bromo-2-trifluoromethylsulfanyl benzene, 1-trifluoromethylthio-2-bromobenzene, 2-bromo-1-trifluoromethylthio benzene, 2-bromophenyl trifluoromethyl sulfide
PubChem CID 2779338
IUPAC Name 1-bromo-2-(trifluoromethylsulfanyl)benzene
SMILES FC(F)(F)SC1=CC=CC=C1Br

Specifications

Boiling Point 92°C to 94°C (20 mmHg)
Refractive Index 1.5125
Quantity 5 g
Formula Weight 257.07
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-(trifluoromethylthio)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.