missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(trifluoromethoxy)benzene, 97%, Thermo Scientific™

Product Code. 11424447
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11424447

Brand: Thermo Scientific Alfa Aesar B20467.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64115-88-4
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.01
MDL Number MFCD00040942
InChI Key ITYCJCVRPBLODP-UHFFFAOYSA-N
Synonym 1-bromo-2-trifluoromethoxy benzene, 2-trifluoromethoxy bromobenzene, 2-bromo trifluoromethoxy benzene, o-trifluoromethoxybromobenzene, o-bromo trifluoromethoxy benzene, 2-bromo-alpha,alpha,alpha-trifluoroanisole, 2-bromophenyl trifluoromethyl ether, trifluoromethoxy bromobenzene, o-bromotrifluoromethoxybenzene, benzene, bromo trifluoromethoxy
PubChem CID 2777266
IUPAC Name 1-bromo-2-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC=C1Br

Specifications

Density 1.632
Boiling Point 158°C to 160°C
Flash Point 60°C (140°F)
Refractive Index 1.4638
Quantity 1 g
UN Number UN1993
Beilstein 1947768
Formula Weight 241.01
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-(trifluoromethoxy)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.