missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-phenoxyethane, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-2-phenoxyethane |
| CAS | 589-10-6 |
| Molecular Formula | C8 H9 Br O |
| Quantity | 2.5 g |
| Synonym | Tamsulosin Impurity 20 |
| InChI Formula | InChI=1S/C8H9BrO/c9-6-7-10-8-4-2-1-3-5-8/h1-5H,6-7H2 |
| SMILES | BrCCOc1ccccc1 |
| IUPAC Name | 2-bromoethoxybenzene |
| Molecular Weight (g/mol) | 201.06 |
| Formula Weight | 199.9837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction