missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-pentyne, 97%, Thermo Scientific Chemicals

Product Code. 11924501 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11924501

Brand: Thermo Scientific Acros 439570100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16400-32-1
Molecular Formula C5H7Br
Molecular Weight (g/mol) 147.02
MDL Number MFCD00236363
InChI Key VDHGRVFJBGRHMD-UHFFFAOYSA-N
Synonym 1-bromo-2-pentyne, 2-pentynyl bromide, 2-pentyne, 1-bromo, 1-bromo-pent-2-yne, 1-bromo-2-pentine, 2-pentyn-1-yl bromide, acmc-1c6g2
PubChem CID 85399
IUPAC Name 1-bromopent-2-yne
SMILES CCC#CCBr

Specifications

Color Yellow
Boiling Point 40°C to 41°C (20.0 mmHg)
Flash Point 36°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.508
Quantity 10 g
Formula Weight 147.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-pentyne
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-2-pentyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.