missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-pentyne, 97%

Product Code. 15470238
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15470238

Brand: Thermo Scientific Alfa Aesar H53433.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16400-32-1 | C5H7Br | 147.02 g/mol

1-Bromo-2-pentyne acts as a reagent for organic and organometallic compounds. It is also employed in the preparation of stereochemically restricted lactone-type analogs of jasmonic acids, 5-oxa-7-epi-jasmonic acid and 5-oxa-jasmonic acid. Further, it is used to prepare of 4,7-decadienal, 4,7-tridecadienal, 5,8-tetradecadienal and 6,9-dodecadienal synthesis. In addition to this, it plays an important role for the synthesis of 5-ethyl-4-methylene-6-phenyl-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-pentyne acts as a reagent for organic and organometallic compounds. It is also employed in the preparation of stereochemically restricted lactone-type analogs of jasmonic acids, 5-oxa-7-epi-jasmonic acid and 5-oxa-jasmonic acid. Further, it is used to prepare of 4,7-decadienal, 4,7-tridecadienal, 5,8-tetradecadienal and 6,9-dodecadienal synthesis. In addition to this, it plays an important role for the synthesis of 5-ethyl-4-methylene-6-phenyl-3a,4,7,7a-tetrahydroisobenzofuran-1,3-dione.

Solubility
Miscible with ether

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16400-32-1
Molecular Formula C5H7Br
Molecular Weight (g/mol) 147.02
MDL Number MFCD00236363
InChI Key VDHGRVFJBGRHMD-UHFFFAOYSA-N
Synonym 1-bromo-2-pentyne, 2-pentynyl bromide, 2-pentyne, 1-bromo, 1-bromo-pent-2-yne, 1-bromo-2-pentine, 2-pentyn-1-yl bromide, acmc-1c6g2
PubChem CID 85399
IUPAC Name 1-bromopent-2-yne
SMILES CCC#CCBr

Specifications

Density 1.438
Boiling Point 93°C to 94°C (113 mmHg)
Flash Point 41°C (106°F)
Linear Formula CH3CH2C≡CCH2Br
Refractive Index 1.498
Quantity 5 g
UN Number UN1993
Solubility Information Miscible with ether
Formula Weight 147.01
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-pentyne
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.