missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-nitrobenzene, 99%

Product Code. 11459183
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459183 1000 g 1000g
11469183 50 g 50g
11479183 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459183 Supplier Thermo Scientific Alfa Aesar Supplier No. A11686.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 577-19-5 | C6H4BrNO2 | 202.007 g/mol

1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2?-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2′-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-19-5
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD00007045
InChI Key ORPVVAKYSXQCJI-UHFFFAOYSA-N
Synonym 2-bromonitrobenzene, o-bromonitrobenzene, o-nitrobromobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, o-nitrophenyl bromide, 2-bromo-1-nitrobenzene, 1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, benzene, bromonitro
PubChem CID 11341
IUPAC Name 1-bromo-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 40°C to 44°C
Boiling Point 261°C
Flash Point 112°C (233°F)
Quantity 1000 g
UN Number UN3459
Beilstein 2044109
Solubility Information Insoluble in water.
Formula Weight 202.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-2-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.