missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-methoxynaphthalene, 97%

Product Code. p-7024377
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493168 5 g 5g
11403178 25 g 25g
11413178 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493168 Supplier Thermo Scientific Alfa Aesar Supplier No. A18926.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3401-47-6 | C11H9BrO | 237.10 g/mol

1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-methoxy-naphthalene is a useful synthetic intermediate. It is used as a reagent to synthesize a catalyst for highly enantioselective aziridination of styrene derivatives. It can also be used to prepare biaryls or biheterocycles by palladium-catalyzed Ullmann coupling.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3401-47-6
Molecular Formula C11H9BrO
Molecular Weight (g/mol) 237.10
MDL Number MFCD00055374
InChI Key XNIGURFWNPLWJM-UHFFFAOYSA-N
Synonym 1-bromo-2-methoxy-naphthalene, naphthalene, 1-bromo-2-methoxy, acmc-1cjky, maybridge3_005134, 2-methoxy-1-bromonaphthalene, 1-bromo-2-methoxy-naphtalene
PubChem CID 72860
IUPAC Name 1-bromo-2-methoxynaphthalene
SMILES COC1=CC=C2C=CC=CC2=C1Br

Specifications

Melting Point 79°C to 80°C
Quantity 5 g
Beilstein 2045600
Solubility Information Slightly soluble in water.
Formula Weight 237.1
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-methoxynaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.