missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper

Product Code. 11480397
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480397 100 g 100g
11460397 5 g 5g
11470397 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480397 Supplier Thermo Scientific Alfa Aesar Supplier No. B21771.22
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 421-70-5 | C2BrF4I | 306.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-70-5
Molecular Formula C2BrF4I
Molecular Weight (g/mol) 306.82
MDL Number MFCD00042136
InChI Key ZYNPYKGTNSXKPI-UHFFFAOYSA-N
Synonym 1-bromo-2-iodotetrafluoroethane, 1-bromotetrafluoro-2-iodoethane, unii-e75xu1n0mr, e75xu1n0mr, ethane, 1-bromo-1,1,2,2-tetrafluoro-2-iodo, ccris 9437, acmc-1amr3, 1-bromo-2-iodoperfluoroethane, 1-iodo-2-bromotetrafluoroethane, 2-iodo-1-bromotetrafluoroethane
PubChem CID 67902
IUPAC Name 1-bromo-1,1,2,2-tetrafluoro-2-iodoethane
SMILES FC(F)(Br)C(F)(F)I

Specifications

Density 1.648
Boiling Point 78°C
Refractive Index 1.433
Quantity 100 g
Beilstein 1688466
Sensitivity Light sensitive
Formula Weight 306.82
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-iodotetrafluoroethane, stabilized with copper

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.