missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-iodobenzene, 99%, stabilized

Product Code. p-3735743 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10323013

Brand: Thermo Scientific Acros 221950010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 583-55-1 | C6H4BrI | 282.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-55-1
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.9
MDL Number MFCD00001030
InChI Key OIRHKGBNGGSCGS-UHFFFAOYSA-N
Synonym 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
PubChem CID 11415
IUPAC Name 1-bromo-2-iodobenzene
SMILES C1=CC=C(C(=C1)Br)I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 9°C to 10°C
Color Yellow
Density 2.2030g/mL
Boiling Point 120°C to 121°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4I
Refractive Index 1.6620 to 1.664
Quantity 1 g
Beilstein 05, 223
Fieser 01,77
Specific Gravity 2.203
Stabilizer copper
Formula Weight 282.9
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-2-iodobenzene, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.