missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-iodobenzene, 98+%, stab. with copper

Product Code. 11448876
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448876 5 g 5g
11458876 25 g 25g
11468876 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448876 Supplier Thermo Scientific Alfa Aesar Supplier No. A17020.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 583-55-1 | C6H4BrI | 282.906 g/mol

2-Bromoiodobenzene is used in the synthesis of diarylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoiodobenzene is used in the synthesis of diarylamines.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-55-1
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.906
MDL Number MFCD00001030
InChI Key OIRHKGBNGGSCGS-UHFFFAOYSA-N
Synonym 2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
PubChem CID 11415
IUPAC Name 1-bromo-2-iodobenzene
SMILES C1=CC=C(C(=C1)Br)I

Specifications

Melting Point 9°C to 10°C
Density 2.256
Boiling Point 257°C
Flash Point >110°C (230°F)
Refractive Index 1.6645
Quantity 5 g
Beilstein 1855309
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 282.91
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2-iodobenzene, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.