missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene, 98%

Product Code. 15498145
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498145 25 g 25g
15425397 1 g 1g
15435397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498145 Supplier Thermo Scientific Alfa Aesar Supplier No. H32650.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 286932-57-8 | C7H3BrF4O | 258.998 g/mol

1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene used as solvents. Mostly employed in chemical processing, extraction and separation of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene used as solvents. Mostly employed in chemical processing, extraction and separation of chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286932-57-8
Molecular Formula C7H3BrF4O
Molecular Weight (g/mol) 258.998
MDL Number MFCD02181186
InChI Key VVVFPEJORKPEST-UHFFFAOYSA-N
Synonym 2-bromo-1-fluoro-4-trifluoromethoxy benzene, 1-bromo-2-fluoro-5-trifluoromethoxy benzene, 3-bromo-4-fluoro trifluoromethoxy benzene, 3-bromo-4-fluorotrifluoroanisole, 2-fluoro-5-trifluoromethoxy-bromobenzene, 3-bromo-4-fluorophenyl trifluoromethyl ether, 3-bromo-4-fluoro-alpha,alpha,alpha-trifluoroanisole, 3-bromo-4-fluorotrifluoromethoxybenzene, pubchem12363, acmc-1cocq
PubChem CID 25067359
IUPAC Name 2-bromo-1-fluoro-4-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1OC(F)(F)F)Br)F

Specifications

Quantity 25 g
Formula Weight 259
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.