missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-fluoro-4-iodobenzene, 97%

Product Code. 11479466
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479466 5 g 5g
11489466 25 g 25g
11499466 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479466 Supplier Thermo Scientific Alfa Aesar Supplier No. A19190.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136434-77-0 | C6H3BrFI | 300.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136434-77-0
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.90
MDL Number MFCD00042577
InChI Key OCODJNASCDFXSR-UHFFFAOYSA-N
Synonym 4-bromo-3-fluoroiodobenzene, 3-fluoro-4-bromo iodobenzene, 1-bromo-2-fluoro-4-iodo-benzene, benzene, 1-bromo-2-fluoro-4-iodo, 3-fluoro-4-bromo-iodobenzene, 1-bromo-2-fluoro-4-i, pubchem3231, acmc-209c5x, 3-fluoro-4-bromoiodobenzene, ksc490o1h
PubChem CID 2725006
IUPAC Name 1-bromo-2-fluoro-4-iodobenzene
SMILES FC1=C(Br)C=CC(I)=C1

Specifications

Melting Point 34°C to 38°C
Boiling Point 80°C (2.5 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 4740251
Sensitivity Light sensitive
Formula Weight 300.9
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-fluoro-4-iodobenzene

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.