missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-ethylhexane, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-2-ethylhexane |
| CAS | 18908-66-2 |
| Molecular Formula | C8H17Br |
| Quantity | 1 g |
| Synonym | 2-Ethyl-1-hexyl Bromide, 2-Ethylhexyl Bromide, 3-(Bromomethyl)heptane |
| InChI Formula | InChI=1S/C8H17Br/c1-3-5-6-8(4-2)7-9/h8H,3-7H2,1-2H3 |
| SMILES | CCCCC(CC)CBr |
| IUPAC Name | 3-(bromomethyl)heptane |
| Molecular Weight (g/mol) | 193.12 |
| Formula Weight | 192.0514 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction