missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethylbutane, 97%

Product Code. 11481420
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481420 5 mL 5mL
11491420 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481420 Supplier Thermo Scientific Alfa Aesar Supplier No. H26445.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3814-34-4 | C6H13Br | 165.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3814-34-4
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.074
MDL Number MFCD00000219
InChI Key KKGUMGWNFARLSL-UHFFFAOYSA-N
Synonym 1-bromo-2-ethylbutane, 3-bromomethyl pentane, 2-ethylbutyl bromide, pentane, 3-bromomethyl, 3-bromomethyl-pentane, butane, 1-bromo-2-ethyl, 2-ethylbutylbromide, bromo-2-ethylbutane, 2-ethyl butyl bromide, 2-ethyl-1-bromobutane
PubChem CID 77432
IUPAC Name 3-(bromomethyl)pentane
SMILES CCC(CC)CBr

Specifications

Density 1.179
Boiling Point 145°C
Flash Point 36°C (96°F)
Odor Sweet
Refractive Index 1.4515
Quantity 5 mL
UN Number UN1993
Formula Weight 165.07
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-ethylbutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.