missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-cyclohexylbenzene, 97%

Product Code. 11314895
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314895 1 g 1g
11324895 5 g 5g
11334895 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314895 Supplier Thermo Scientific Alfa Aesar Supplier No. L18059.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59734-92-8 | C12H15Br | 239.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59734-92-8
Molecular Formula C12H15Br
Molecular Weight (g/mol) 239.16
MDL Number MFCD02093987
InChI Key BXMDAGGQRIDSPA-UHFFFAOYSA-N
Synonym 1-bromo-2-cyclohexyl-benzene, benzene, 1-bromo-2-cyclohexyl, acmc-20aoxa, l-bromo-2-cyclohexylbenzene, 2-bromo-1-cyclohexylbenzene
PubChem CID 10944502
IUPAC Name 1-bromo-2-cyclohexylbenzene
SMILES BrC1=CC=CC=C1C1CCCCC1

Specifications

Density 1.295
Boiling Point 79°C to 83°C (0.01 mmHg)
Refractive Index 1.5605
Quantity 1 g
Beilstein 2365764
Formula Weight 239.16
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-cyclohexylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.