missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloroethane, 98%

Product Code. 11489473
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11489473

Brand: Thermo Scientific Alfa Aesar A12349.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-04-0 | C2H4BrCl | 143.408 g/mol

1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

Solubility
Soluble in water ( 7 g/L (20°C). Soluble in ethanol, ethyl ether, chloroform.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-04-0
Molecular Formula C2H4BrCl
Molecular Weight (g/mol) 143.408
MDL Number MFCD00000962
InChI Key IBYHHJPAARCAIE-UHFFFAOYSA-N
Synonym ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
PubChem CID 7849
ChEBI CHEBI:19032
IUPAC Name 1-bromo-2-chloroethane
SMILES C(CBr)Cl

Specifications

Melting Point -18°C
Density 1.726
Boiling Point 106°C to 107°C
Flash Point None
Odor Sweet
Linear Formula ClCH2CH2Br
Refractive Index 1.49
Quantity 2500 g
UN Number UN2810
Beilstein 605265
Sensitivity Light sensitive
Solubility Information Soluble in water ( 7g/L (20°C). Soluble in ethanol,ethyl ether,chloroform.
Formula Weight 143.42
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-chloroethane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.