missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloroethane, 98%

Product Code. 11479473
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479473 500 g 500g
11469473 100 g 100g
11489473 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479473 Supplier Thermo Scientific Alfa Aesar Supplier No. A12349.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-04-0 | C2H4BrCl | 143.408 g/mol

1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

Solubility
Soluble in water ( 7 g/L (20°C). Soluble in ethanol, ethyl ether, chloroform.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-04-0
Molecular Formula C2H4BrCl
Molecular Weight (g/mol) 143.408
MDL Number MFCD00000962
InChI Key IBYHHJPAARCAIE-UHFFFAOYSA-N
Synonym ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
PubChem CID 7849
ChEBI CHEBI:19032
IUPAC Name 1-bromo-2-chloroethane
SMILES C(CBr)Cl

Specifications

Melting Point -18°C
Density 1.726
Boiling Point 106°C to 107°C
Flash Point None
Odor Sweet
Linear Formula ClCH2CH2Br
Refractive Index 1.49
Quantity 500 g
UN Number UN2810
Beilstein 605265
Sensitivity Light sensitive
Solubility Information Soluble in water ( 7g/L (20°C). Soluble in ethanol,ethyl ether,chloroform.
Formula Weight 143.42
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-chloroethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-2-chloroethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 1-Bromo-2-chloroethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.