missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chlorobenzene, 98+%

Product Code. 11459353
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459353 50 g 50g
11469353 250 g 250g
11449353 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459353 Supplier Thermo Scientific Alfa Aesar Supplier No. A11782.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 694-80-4 | C6H4BrCl | 191.452 g/mol

1-Bromo-2-chlorobenzene finds application in Gas Chromatography and Liquid Chromatography analysis. It is also used as an intermediate in pharmaceutical and synthetic chemistry. Further, it acts as a reagent in the preparation of arylpiperidines and aryltetrahydropyridines as 5-HT2C agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-chlorobenzene finds application in Gas Chromatography and Liquid Chromatography analysis. It is also used as an intermediate in pharmaceutical and synthetic chemistry. Further, it acts as a reagent in the preparation of arylpiperidines and aryltetrahydropyridines as 5-HT2C agonists.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-80-4
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.452
MDL Number MFCD00000532
InChI Key QBELEDRHMPMKHP-UHFFFAOYSA-N
Synonym 2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene
PubChem CID 12754
IUPAC Name 1-bromo-2-chlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Br

Specifications

Melting Point -13°C
Density 1.649
Quantity 50 g
Refractive Index 1.5815
Beilstein 1855303
Boiling Point 203°C to 205°C
Solubility Information Immiscible with water.
Formula Weight 191.46
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2-chlorobenzene
compliance-icons
Product Identifier
  • 1-Bromo-2-chlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Bromo-2-chlorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.