missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals

Product Code. 11477216
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11477216

Brand: Thermo Scientific Alfa Aesar B25538.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110407-59-5
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00142587
InChI Key LEFQPBAWVJEIJS-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobromobenzene, 4-bromo-3-chloro-1-fluorobenzene, 1-bromo-2-chloro-4-fluoro-benzene, 4-bromo-3-chlorofluorobenzene, 2-bromo-5-fluorochlorobenzene, benzene, 1-bromo-2-chloro-4-fluoro, pubchem3755, acmc-1bnv0, 2-chloro-4-fluorobromobezene, ksc494m7f
PubChem CID 2733406
IUPAC Name 1-bromo-2-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)Br

Specifications

Refractive Index 1.552
Quantity 5 g
Formula Weight 209.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-chloro-4-fluorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.