missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-butyne, 98%

Product Code. 11374586
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374586 5 g 5g
11384586 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374586 Supplier Thermo Scientific Alfa Aesar Supplier No. L20134.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3355-28-0 | C4H5Br | 132.988 g/mol

1-Bromo-2-butyne is used in the preparation of six to eight annulated ring compounds in reaction with indoles and pseudopterane (±)-Kallolide B, which is a marine natural product. Further, it acts as a precursor in the preparation of axially chiral teranyl compounds, alkylation of L-tryptophan methyl ester, 4-butynyloxybenzene sulfonyl chloride and mono-propargylated diene derivative. In addition to this, it is also used in the synthesis of isopropylbut-2-ynylamine, allenylcyclobutanol derivatives, allyl-[4-(but-2-ynyloxy)phenyl]sulfane, allenylindium and axially chiral teranyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-butyne is used in the preparation of six to eight annulated ring compounds in reaction with indoles and pseudopterane (+/-)-Kallolide B, which is a marine natural product. Further, it acts as a precursor in the preparation of axially chiral teranyl compounds, alkylation of L-tryptophan methyl ester, 4-butynyloxybenzene sulfonyl chloride and mono-propargylated diene derivative. In addition to this, it is also used in the synthesis of isopropylbut-2-ynylamine, allenylcyclobutanol derivatives, allyl-[4-(but-2-ynyloxy)phenyl]sulfane, allenylindium and axially chiral teranyl compounds.

Solubility
Miscible with acetonitrile.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3355-28-0
Molecular Formula C4H5Br
Molecular Weight (g/mol) 132.988
MDL Number MFCD00190233
InChI Key LNNXOEHOXSYWLD-UHFFFAOYSA-N
Synonym 1-bromo-2-butyne, 2-butyne, 1-bromo, 2-butynyl bromide, 1-bromo-but-2-yne, bromo-2-butyne, 1-bromo2-butyne, but-2-ynylbromide, l-bromo-2-butine, 1-bromo-2-butine, 4-bromo-2-butyne
PubChem CID 2756862
IUPAC Name 1-bromobut-2-yne
SMILES CC#CCBr

Specifications

Density 1.519
Boiling Point 123°C to 125°C
Flash Point 36°C (96°F)
Refractive Index 1.508
Quantity 5 g
UN Number UN1993
Beilstein 605306
Sensitivity Air Sensitive
Solubility Information Miscible with acetonitrile.
Formula Weight 132.99
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-butyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.