missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals

Product Code. 10119612 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10119612 1 g Glass bottle 1g
10495993 5 g Glass bottle 5g
10781725 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10119612 Supplier Thermo Scientific Acros Supplier No. 222450010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 816-40-0
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151
MDL Number MFCD00000207
InChI Key CCXQVBSQUQCEEO-UHFFFAOYSA-N
Synonym 1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one
PubChem CID 13156
IUPAC Name 1-bromobutan-2-one
SMILES CCC(=O)CBr

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4790g/mL
Boiling Point 105°C (150.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C2H5COCH2Br
Refractive Index 1.4650 to 1.469
Quantity 1 g
Beilstein 01,67
Specific Gravity 1.479
Stabilizer approx. 0.5% CaCO3
Formula Weight 151
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-butanone, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-2-butanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.