missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,6-dichlorobenzene, 99%, Thermo Scientific™

Product Code. 10114190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10114190 5 g Glass bottle 5g
10092090 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10114190 Supplier Thermo Scientific Acros Supplier No. 171040050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19393-92-1
MDL Number MFCD00000574
InChI Key UWOIDOQAQPUVOH-UHFFFAOYSA-N
Synonym 1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
PubChem CID 29568
IUPAC Name 2-bromo-1,3-dichlorobenzene
SMILES C1=CC(=C(C(=C1)Cl)Br)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 64.0°C to 67.0°C
Color White to Yellow
Boiling Point 242.0°C (765.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H3BrCl2
Linear Formula BrC6H3Cl2
Quantity 5 g
Beilstein 05, 210
Molecular Weight (g/mol) 225.9
Formula Weight 225.9
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-Bromo-2, 6-dichlorobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromo-2,6-dichlorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.