missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dinitrobenzene, MP Biomedicals

Product Code. 11431056 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431056 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11431056 Supplier MP Biomedicals Supplier No. 0520234410
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 584-48-5
Molecular Formula C6H3BrN2O4
Quantity 10 g
Synonym 2,4-dinitrobromobenzene, 4-bromo-1,3-dinitrobenzene, o,p-dinitrophenyl bromide, 2,4-dinitrophenyl bromide, dnbb, benzene, 1-bromo-2,4-dinitro, 2, 4-dinitrobromobenzene, 1-bromo-2,4-dinitrobenze, unii-4ms3ftw380, ccris 1801
InChI Key PBOPJYORIDJAFE-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Br
IUPAC Name 1-bromo-2,4-dinitrobenzene
Molecular Weight (g/mol) 247.004
PubChem CID 11441
ChEBI CHEBI:58999
Formula Weight 247.01
Chemical Name or Material 1-Bromo-2, 4-dinitrobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.