missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-dinitrobenzene, 98%

Product Code. 11469917
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11469917

Brand: Thermo Scientific Alfa Aesar B22867.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 584-48-5 | C6H3BrN2O4 | 247.004 g/mol

1-Bromo-2,4-dinitrobenzene has been used in the preparation of 2,4-dinitrophenol via treatment with KO2-crown ether complex in benzene. It is used as substrate in protein determination and glutathione S-transferase (GST) assay of chicken and rat prostaglandin D2 synthase (PGDS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2,4-dinitrobenzene has been used in the preparation of 2,4-dinitrophenol via treatment with KO2-crown ether complex in benzene. It is used as substrate in protein determination and glutathione S-transferase (GST) assay of chicken and rat prostaglandin D2 synthase (PGDS).

Solubility
Soluble in water (partly miscible).

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-48-5
Molecular Formula C6H3BrN2O4
Molecular Weight (g/mol) 247.004
MDL Number MFCD00041873
InChI Key PBOPJYORIDJAFE-UHFFFAOYSA-N
PubChem CID 11441
ChEBI CHEBI:58999
IUPAC Name 1-bromo-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Br

Specifications

Melting Point 70°C to 73°C
Quantity 100 g
UN Number UN3459
Synonym 2,4-dinitrobromobenzene, 4-bromo-1,3-dinitrobenzene, o,p-dinitrophenyl bromide, 2,4-dinitrophenyl bromide, dnbb, benzene, 1-bromo-2,4-dinitro, 2, 4-dinitrobromobenzene, 1-bromo-2,4-dinitrobenze, unii-4ms3ftw380, ccris 1801
Solubility Information Soluble in water (partly miscible).
Formula Weight 247.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,4-dinitrobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.