missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4-bis(trifluoromethyl)benzene, 97%, Thermo Scientific™

Product Code. 11420498
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11420498

Brand: Thermo Scientific Alfa Aesar B24630.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-75-3
Molecular Formula C8H3BrF6
Molecular Weight (g/mol) 293.01
MDL Number MFCD00074904
InChI Key QDEJWLIKRLJYEK-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl bromobenzene, 1-bromo-2,4-bis trifluoromethyl benzene, 4-bromo-1,3-bis trifluoromethyl benzene, 1,3-bis trifluoromethyl-4-bromobenzene, 2,4-ditrifluoromethylbromobenzene, benzene, 1-bromo-2,4-bis trifluoromethyl, 2,4-bis-trifluoromethylbromobenzene, fxffr be exfff, pubchem8494
PubChem CID 136164
IUPAC Name 1-bromo-2,4-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=C(Br)C(=C1)C(F)(F)F

Specifications

Density 1.738
Boiling Point 163°C
Flash Point >110°C (230°F)
Refractive Index 1.437
Quantity 1 g
Beilstein 6208648
Formula Weight 293.01
Percent Purity 97%
Chemical Name or Material 1-Bromo-2,4-bis(trifluoromethyl)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.