missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4,5-trifluorobenzene, 98%, Thermo Scientific™

Product Code. 10093240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Glass bottle; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10093240 Glass bottle; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10093240 Supplier Thermo Scientific Acros Supplier No. 271160050
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 327-52-6
CAS Min % 98.5
CAS Max % 100.0
Density 1.8020g/mL
Boiling Point 143.0°C to 144.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H2BrF3
Linear Formula BrC6H2F3
Refractive Index 1.4850 to 1.4870
MDL Number MFCD00000306
Quantity 5g
Beilstein 05, 679
Specific Gravity 1.802
Synonym 2,4,5-trifluorobromobenzene, 2-bromo-1,4,5-trifluorobenzene, benzene, 1-bromo-2,4,5-trifluoro, 1-bromo-2,4,5-trifluoro-benzene, paragos 390205, 5-bromo-1,2,4-trifluorobenzene, fr bf df ee, pubchem2306, acmc-1cs6j, ksc224g3h
Solubility Information Solubility in water: insoluble.
InChI Key DVTULTINXNWGJY-UHFFFAOYSA-N
SMILES C1=C(C(=CC(=C1F)Br)F)F
IUPAC Name 1-bromo-2,4,5-trifluorobenzene
Molecular Weight (g/mol) 210.98
PubChem CID 67597
Formula Weight 210.98
Percent Purity 98%
Chemical Name or Material 1-Bromo-2, 4, 5-trifluorobenzene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.