missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,4,5-trifluorobenzene, 98+%, Thermo Scientific™

Product Code. 11473974
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473974 1 g 1g
11483974 5 g 5g
11493974 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473974 Supplier Thermo Scientific Alfa Aesar Supplier No. A11962.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Bromo-2,4,5-trifluorobenzene is the intermediate of liquid crystals. The derivatives of this compound can be used as intermediates for the synthesis of qunolone antibacterial. It is employed in the synthesis of 3-ethenyl, 3-ethynyl, 3-aryl, and 3-cyclopropyl-2, 4, 5-trifluorobenzoic acids from 1-bromo-2, 4, 5-trifluorobenzene and 2, 4, 5-trifluoro-3-hydroxybenzoic acid.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-52-6
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.981
MDL Number MFCD00000306
InChI Key DVTULTINXNWGJY-UHFFFAOYSA-N
Synonym 2,4,5-trifluorobromobenzene, 2-bromo-1,4,5-trifluorobenzene, benzene, 1-bromo-2,4,5-trifluoro, 1-bromo-2,4,5-trifluoro-benzene, paragos 390205, 5-bromo-1,2,4-trifluorobenzene, fr bf df ee, pubchem2306, acmc-1cs6j, ksc224g3h
PubChem CID 67597
IUPAC Name 1-bromo-2,4,5-trifluorobenzene
SMILES C1=C(C(=CC(=C1F)Br)F)F

Specifications

Melting Point -19°C
Density 1.806
Boiling Point 143°C to 144°C
Flash Point 55°C (131°F)
Refractive Index 1.486
Quantity 1 g
UN Number UN1993
Beilstein 1636588
Solubility Information Insoluble in water.
Formula Weight 210.98
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2,4,5-trifluorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.