missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,5-trifluorobenzene, 98%, Thermo Scientific™

Product Code. 11490441
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490441 5 g 5g
11480441 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490441 Supplier Thermo Scientific Alfa Aesar Supplier No. B20000.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133739-70-5
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.98
MDL Number MFCD00012232
InChI Key XSMLLZPSNLQCQU-UHFFFAOYSA-N
Synonym 2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
PubChem CID 2736399
IUPAC Name 1-bromo-2,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(F)C(Br)=C1

Specifications

Density 1.777
Boiling Point 143°C
Flash Point 54°C (129°F)
Refractive Index 1.485
Quantity 5 g
UN Number UN1993
Beilstein 7369071
Formula Weight 210.98
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,3,5-trifluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.