missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,5-trifluorobenzene, 98%, Thermo Scientific™

Product Code. p-4323174 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11954551

Brand: Thermo Scientific Acros 443840010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 133739-70-5
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.98
MDL Number MFCD00012232
InChI Key XSMLLZPSNLQCQU-UHFFFAOYSA-N
Synonym 2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
PubChem CID 2736399
IUPAC Name 1-bromo-2,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(F)C(Br)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 143°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 210.98
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2, 3, 5-trifluorobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-2,3,5-trifluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.