missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,5,6-tetramethylbenzene, 99%

Product Code. 11391277
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11391277

Brand: Thermo Scientific Alfa Aesar L01465.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1646-53-3 | C10H13Br | 213.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1646-53-3
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00000070
InChI Key WJKBPTLQJXKEHC-UHFFFAOYSA-N
Synonym bromodurene, 3-bromodurene, 1-bromo-2,3,5,6-tetramethylbenzene, benzene, 3-bromo-1,2,4,5-tetramethyl, bromdurol, duryl bromide, bromodurene, pract., acmc-1bz04, 2,3,5,6-tetramethylbromobenzene, 1-bromo-2,5,6-tetramethylbenzene
PubChem CID 74240
IUPAC Name 3-bromo-1,2,4,5-tetramethylbenzene
SMILES CC1=CC(=C(C(=C1C)Br)C)C

Specifications

Melting Point 58°C to 62°C
Boiling Point 115°C to 120°C (5 mmHg)
Quantity 5 g
Beilstein 2207158
Formula Weight 213.12
Percent Purity 99%
Chemical Name or Material 1-Bromo-2,3,5,6-tetramethylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.