missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(2-methoxyethoxy)ethane, tech. 90%, stab. with sodium carbonate

Product Code. 11347435
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347435 25 g 25g
11337435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347435 Supplier Thermo Scientific Alfa Aesar Supplier No. L05406.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54149-17-6 | C5H11BrO2 | 183.05 g/mol

1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-(2-methoxyethoxy)ethane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store at 4°C. Protect from heat. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54149-17-6
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000238
InChI Key HUXJXNSHCKHFIL-UHFFFAOYSA-N
Synonym 1-bromo-2-2-methoxyethoxy ethane, 1-2-bromoethoxy-2-methoxyethane, 1-bromo-3,6-dioxaheptane, ethane, 1-2-bromoethoxy-2-methoxy, 2-2-methoxyethoxy ethyl bromide, 1-2-bromoethoxy-2-methoxy-ethane, 1-bromo-2-2-methoxy-ethoxy ethane, mpeg2-br, acmc-1alws, 7-bromo-2,5-dioxaheptane
PubChem CID 123532
IUPAC Name 1-(2-bromoethoxy)-2-methoxyethane
SMILES COCCOCCBr

Specifications

Density 1.347
Boiling Point 156°C
Flash Point 73°C (163°F)
Refractive Index 1.455
Quantity 25 g
Beilstein 2069579
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 183.05
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Bromo-2-(2-methoxyethoxy)ethane, stabilized with sodium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.