missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,2-dimethoxypropane, 98%, Thermo Scientific Chemicals

Catalog Number 11449043
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
126-38-5
C5H11BrO2
183.045
MFCD00000208
SGTITUFGCGGICE-UHFFFAOYSA-N
propane,1-bromo-2,2-dimethoxy, propane, 1-bromo-2,2-dimethoxy, bromoacetone dimethyl acetal, bromo-2,2dimethoxypropane, bromo-2,2-dimethoxypropane, 1-bromo-2,2-dimethoxypropan, 1-bromo-2,2-dimethoxyprpopane, 1-bromo-2,2-dimethoxy propane, 1-bromo-2,2-dimethoxy-propane, bromoacetone dimethyl ketal
67175
1-bromo-2,2-dimethoxypropane
CC(CBr)(OC)OC
This item is not returnable. View return policy
126-38-5
C5H11BrO2
183.045
MFCD00000208
SGTITUFGCGGICE-UHFFFAOYSA-N
propane,1-bromo-2,2-dimethoxy, propane, 1-bromo-2,2-dimethoxy, bromoacetone dimethyl acetal, bromo-2,2dimethoxypropane, bromo-2,2-dimethoxypropane, 1-bromo-2,2-dimethoxypropan, 1-bromo-2,2-dimethoxyprpopane, 1-bromo-2,2-dimethoxy propane, 1-bromo-2,2-dimethoxy-propane, bromoacetone dimethyl ketal
67175
1-bromo-2,2-dimethoxypropane
CC(CBr)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.354
Boiling Point 83°C to 87°C (80 mmHg)
Flash Point 40°C (104°F)
Refractive Index 1.447
Quantity 10 g
UN Number UN3271
Beilstein 2036095
Sensitivity Moisture Sensitive
Formula Weight 183.05
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,2-dimethoxypropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'