missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,2-dimethoxypropane, 98%

Product Code. 11469043
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469043 250 g 250g
11449043 10 g 10g
11459043 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469043 Supplier Thermo Scientific Alfa Aesar Supplier No. A11139.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126-38-5 | C5H11BrO2 | 183.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-38-5
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.045
MDL Number MFCD00000208
InChI Key SGTITUFGCGGICE-UHFFFAOYSA-N
Synonym propane,1-bromo-2,2-dimethoxy, propane, 1-bromo-2,2-dimethoxy, bromoacetone dimethyl acetal, bromo-2,2dimethoxypropane, bromo-2,2-dimethoxypropane, 1-bromo-2,2-dimethoxypropan, 1-bromo-2,2-dimethoxyprpopane, 1-bromo-2,2-dimethoxy propane, 1-bromo-2,2-dimethoxy-propane, bromoacetone dimethyl ketal
PubChem CID 67175
IUPAC Name 1-bromo-2,2-dimethoxypropane
SMILES CC(CBr)(OC)OC

Specifications

Density 1.354
Boiling Point 83°C to 87°C (80 mmHg)
Flash Point 40°C (104°F)
Refractive Index 1.447
Quantity 250 g
UN Number UN3271
Beilstein 2036095
Sensitivity Moisture Sensitive
Formula Weight 183.05
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,2-dimethoxypropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.